Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Methyl Lys27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244208
Description
Specifically detects Histone H3 (Methyl Lys27) in C. elegans, Human, Mouse, Rat samples, and it is validated for Western Blotting
Histone H3 Polyclonal specifically detects Histone H3 in Human samples. It is validated for Western Blot, Dot Blot.Specifications
Histone H3 (Methyl Lys27) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
50 μg | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Human, Mouse, Rat, C. elegans | |
Purified |
Western Blot, Dot Blot | |
0.99 mg/mL | |
Western Blot 1:1000, Dot Blot | |
Q71DI3 | |
H3C14 | |
Histone H3 [Methyl Lys27] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide surrounding the K27 me3 site of human Histone H3. (Uniprot: Q71DI3) | |
Protein A purified | |
RUO | |
Primary | |
This antibody recognizes the Me1, Me2, Me3, and acetylated form of Lys27 for human Histone H3. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction