Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, Monomethyl Lys56 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $402.00

Specifications

Antigen Histone H3 (Monomethyl Lys56)
Concentration 0.63 mg/mL
Dilution Western Blot 1 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, In vitro assay, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, In vitro Assay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB211341SS
SDP
View Documents
Novus Biologicals
NB211341SS
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NB211341
SDP
View Documents
Novus Biologicals
NB211341
0.05 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, In vitro assay, Dot Blot.
Specifications

Specifications

Histone H3 (Monomethyl Lys56)
Western Blot 1 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, In vitro assay, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells
Polyclonal
Rabbit
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
PBS and 30% Glycerol with 0.05% Sodium Azide
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K56, H3K56Me1, HIST2H3C, histone H3.3, MGC87782, MGC87783
H3C14
IgG
Affinity Purified
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
0.63 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, In vitro Assay
Unconjugated
RUO
Human, Mouse, C. elegans, Other
Q71DI3, P84228
126961
This Histone H3 [Monomethyl Lys56] antibody was raised against synthetic monomethylated peptide surrounding lysine 56 of human Histone H3 [Swiss Prot Q71DI3].
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
15 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.