Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 126961 |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized. Reconstitute in diH20. |
| For Use With (Application) | Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay |
| Product Type | Histone H3 |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | ARTKQTAR[KMe1]STGGKAPRKQLA(K-Ahx- Biot)-amide |
| Termination | Biotin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
