Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, p Ser28 Antibody (117C826) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233237
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human, Fish samples. It is validated for Western Blot.Specifications
| Histone H3 (p Ser28) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1-2 ug/ml | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3S28p, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Mouse | |
| 15 kDa | |
| 0.1 mg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Human, Fish, C. elegans, Monkey | |
| Purified |
| Western Blot | |
| 117C826 | |
| Unconjugated | |
| Q71D13 | |
| H3C14 | |
| This Histone H3 [p Ser28] antibody (117C826) - Azide and BSA Free was raised against a synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK of human Histone H3. The immunogen sequence (CARKS*APATGGVKK) is conserved across mammalian and invertebrates, as well as plants. | |
| Protein G purified | |
| RUO | |
| Primary | |
| In anti-Fas mAb treated Jurkat, an 18 kDa band is observed using Histone H3 [p Ser28] antibody (117C826). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction