Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, p Ser28 Antibody (117C826) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen Histone H3 (p Ser28)
Clone 117C826
Concentration 0.5 mg/mL
Dilution Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 22279221), Immunohistochemistry-Paraffin reported in scientific literature (PMID 35897708)
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056595T
SDP
View Documents
Novus Biologicals
NB10056595SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056595
SDP
View Documents
Novus Biologicals
NB10056595
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Histone H3 Monoclonal specifically detects Histone H3 in Human, Fish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Histone H3 (p Ser28)
0.5 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Q71DI3
126961
This Histone H3 [p Ser28] antibody (117C826) was raised against a synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK of human Histone H3. The immunogen sequence is conserved across mammalian and invertebrates, as well as plants.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
15 kDa
117C826
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 22279221), Immunohistochemistry-Paraffin reported in scientific literature (PMID 35897708)
Monoclonal
Purified
RUO
Human
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3S28p, HIST2H3C, histone H3.3, MGC87782, MGC87783
H3C14
IgG1
Protein G purified
In anti-Fas mAb treated Jurkat, an 18 kDa band is observed using Histone H3 [p Ser28] antibody (117C826).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.