Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, p Thr6, Monomethyl Lys9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $402.00
Specifications
| Antigen | Histone H3 (p Thr6, Monomethyl Lys9) |
|---|---|
| Concentration | 0.85 mg/mL |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence 1:200, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB211052SS
![]() |
Novus Biologicals
NB211052SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB211052
![]() |
Novus Biologicals
NB211052 |
0.05 mg |
Each for $402.00
|
|
|||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
| Histone H3 (p Thr6, Monomethyl Lys9) | |
| Western Blot, Immunocytochemistry/Immunofluorescence 1:200, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells | |
| Polyclonal | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me1, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| H3C14 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. |
| 0.85 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, C. elegans | |
| Q71DI3, P84228 | |
| 126961 | |
| This Histone H3 [p Thr6, Monomethyl Lys9] antibody was raised against synthetic monomethylated/phosphorylated peptide surrounding Lysine 9/ Threonine 6 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title