Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 (phospho S10) Mouse anti-Human, Mouse, Clone: 44AT1032.72, Abnova™

Catalog No. 89032548 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-548 Supplier Abnova Corporation Supplier No. MAB15162

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic phosphopeptide of Histone H3.

  • High specificity
  • High reactivity

Specifications

Antigen Histone H3 (phospho S10)
Applications Western Blot
Classification Monoclonal
Clone 44AT1032.72
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide)
Gene Accession No. P84243
Host Species Mouse
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to amino acids surrounding S10 of human Histone H3.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at −20°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.