Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 (phospho S28), Rabbit anti-Human, PE, Clone: D6, Abnova™

Catalog No. 89335753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-753 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-753 Supplier Abnova Corporation Supplier No. MAB18919
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen Histone H3 (phospho S28)
Applications Flow Cytometry
Classification Monoclonal
Clone D6
Conjugate PE
Dilution Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S28 of human Histone H3.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.