Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Rabbit anti-Human, Clone: PHH3/471R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
| Antigen | Histone H3 |
|---|---|
| Clone | PHH3/471R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Histone H3 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 1 mM PBS with 0.05% BSA | |
| 126961 | |
| IgG κ | |
| Protein A or G purified |
| PHH3/471R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Synthetic peptide corresponding to Phosphohistone H3 (PHH3) residues within aa1-100 (phospho S10) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title