Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Rabbit anti-Human, KHL, Polyclonal, MilliporeSigma™

Catalog No. ABE288210MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE288210MI 100 μL
ABE288225MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE288210MI Supplier MilliporeSigma Supplier No. ABE2882100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 Polyclonal antibody specifically detects Histone H3 in Human samples. It is validated for Dot Blot, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate KLH
Gene Accession No. NP_003520
Gene Alias Histone H3/a;Histone H3/b;Histone H3/c;Histone H3/d;Histone H3/f;Histone H3/h;Histone H3/I;Histone H3/j;Histone H3/k;Histone H3/l
Gene Symbols HIST1H3A, H3FA, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3E, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 11 amino acids from the N-terminal region of Human Histone H3 with lysine 14 substituted with methionine.
Molecular Weight of Antigen ∼19 kDa observed; 15.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μL
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects Histone H3 with K14M substitution.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.