Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Rabbit anti-Human, Mouse, Rat, Clone: 7W5T2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $516.00
Specifications
Antigen | Histone H3 |
---|---|
Clone | 7W5T2 |
Applications | ChIP Assay, Dot Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Dot BlotSpecifications
Histone H3 | |
ChIP Assay, Dot Blot | |
Unconjugated | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
126961 | |
IgG | |
Affinity purified |
7W5T2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
A synthetic trimethylated peptide around K36 of human Histone H3 (NP_003520.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title