Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 (tri methyl K37) Polyclonal Antibody, Biotin, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3768RBIOTIN
This item is not returnable.
View return policy
Description
Histone H3 (tri methyl K37) Polyclonal antibody specifically detects Histone H3 (tri methyl K37) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western BlotSpecifications
| Histone H3 (tri methyl K37) | |
| Polyclonal | |
| Biotin | |
| H3C15 | |
| H3 histone; H3/b; H3/d; H3/f; H3/j; Histone 3; Histone H3; methyl Histone 3; methyl Histone H3 | |
| KLH conjugated synthetic methylpeptide derived from human Histone H3 around methylation site of K37 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| P68431, P68433, P84228, Q71DI3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 126961, 260423, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 8968, 97114 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction