Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, Trimethyl Lys27 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP259206

Catalog No. NBP259206


Only null left
Add to Cart

Description

Description

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, A. thaliana, Drosophila, Parasite, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot, Chromatin Immunoprecipitation Sequencing, Knockdown Validated.
Specifications

Specifications

Histone H3 (Trimethyl Lys27)
Polyclonal
Western Blot 1:1000, ELISA 1:200, Immunocytochemistry/Immunofluorescence 1:500, Dot Blot 1:20000, Chromatin Immunoprecipitation (ChIP) 1 ug/IP, Chromatin Immunoprecipitation Sequencing, Knockdown Validated
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K27Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783
Rabbit
15 kDa
50 μg
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
126961
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
H3C14
The exact sequence of the immunogen to this Histone H3 [Trimethyl Lys27] antibody is proprietary.
Affinity Purified
RUO
Primary
Human, Mouse, Rat, A. thaliana, Drosophila, Parasite, Zebrafish
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only