Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Trimethyl Lys37 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$203.51 - $394.26
Specifications
| Antigen | Histone H3 (Trimethyl Lys37) |
|---|---|
| Concentration | 0.76 mg/mL |
| Dilution | Western Blot 1ug/ml, Dot Blot 1:1000, ICC/IF (Negative) |
| Applications | Western Blot, Dot Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB211273SS
![]() |
Novus Biologicals
NB211273SS |
0.025 mg |
Each for $203.51
|
|
|||||
NB211273
![]() |
Novus Biologicals
NB211273 |
0.05 mg |
Each for $394.26
|
|
|||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Dot Blot, ICC/IF (Negative).Specifications
| Histone H3 (Trimethyl Lys37) | |
| Western Blot 1ug/ml, Dot Blot 1:1000, ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| 126961 | |
| This Histone H3 [Trimethyl Lys37] antibody was raised against synthetic trimethylated peptide surrounding Lysine 37 of human Histone H3 [Swiss Prot# Q71DI3]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 15 kDa |
| 0.76 mg/mL | |
| Western Blot, Dot Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse, C. elegans | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K37Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| H3C14 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title