Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Trimethyl Lys9 Antibody (6F12-H4), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130141MFV450
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammalia, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)Specifications
Histone H3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, C. elegans | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
6F12-H4 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
This Histone H3 [Trimethyl Lys9] antibody (6F12-H4) was raised against a synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243] | |
0.1 mL | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction