Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Trimethyl Lys9 Antibody (6F12-H4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 19 publications
Supplier: Novus Biologicals NBP130141SS
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammal, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Histone H3 (Trimethyl Lys9) | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000. Use reported in scientific literature (PMID 23872316), Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 28270554), Immunohistochemistry-Paraffin 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Mouse | |
| 15 kDa | |
| 0.025 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Human, Mouse, Rat, Pig, C. elegans, Drosophila, Invertebrate, Mammalia, Yeast | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 6F12-H4 | |
| Unconjugated | |
| P84243, P84244 | |
| H3C14 | |
| This Histone H3 [Trimethyl Lys9] antibody (6F12-H4) was raised against a synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243] | |
| Protein G purified | |
| RUO | |
| Primary | |
| All mammals including human, mouse and rat, Drosophila, C. elegans, yeast. Invertebrate/Blattella germanica (German cockroach) reactivity reported in scientific literature (PMID: 23872316). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction