Learn More
Description
Specifications
Specifications
| Antigen | Histone H3 (Trimethyl Lys9) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 6F12-H4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000. Use reported in scientific literature (PMID 23872316), Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 28270554), Immunohistochemistry-Paraffin 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Gene Accession No. | P84243, P84244 |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbols | H3C14 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
