Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3. X/Y Rat anti-Human, Clone: 8H6-2111, MilliporeSigma™

Catalog No. MABE243IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE243IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE243IMI Supplier MilliporeSigma Supplier No. MABE243I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Histone H3. X/Y Antibody, clone 8H6-2111 is a highly specific rat monoclonal antibody that targets Histone H3. X/Y and has been tested in Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3. X/Y
Applications Immunocytochemistry
Classification Monoclonal
Clone 8H6-2111
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols H3.X/Y
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to a sequence of 12 amino acids from the N-terminal region of human Histone H3.X/Y.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 8H6-2111 specifically detects X and Y variants of Histone H3 in primates. It does not recognize HA-H3.1, -H3.2, or -H3.3. This clone targets an epitope of 12 amino acids in the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.