Learn More
Histone H3ac (pan-acetyl) Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H3ac (pan-acetyl) |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Host Species | Rabbit |
| Immunogen | This Histone H3 acetyl antibody was raised against a peptide including acetyl-lysines contained in the N-terminal tail of histone H3. |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.