Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3K27ac Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$156.65 - $638.65

Specifications

Antigen Histone H3K27ac
Concentration Lot-Specific
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2635
SDP
Encompass
View Documents
Active Motif
39134
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-199-2636
SDP
Encompass
View Documents
Active Motif
39034
50 μg
Each for $367.53
Only null left
Add to Cart
 
50-199-2634
SDP
Encompass
View Documents
Active Motif
39133
100 μg
Each for $638.65
Only null left
Add to Cart
 
Description

Description

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Specifications

Specifications

Histone H3K27ac
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human
P68431, P84243, Q6NXT2, Q71DI3
126961, 3020, 3021, 333932, 440093, 8350, 8357, 8358, 8968
This Histone H3 acetyl Lys27 antibody was raised against a peptide including acetyl-lysine 27 of histone H3.
Primary
-20°C, Avoid Freeze/Thaw Cycles
H3-3A
Lot-Specific
Polyclonal
Liquid
RUO
10mM sodium phosphate with 150mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5
Acetyl H3; Acetyl Histone H3; Acetyl-H3 K27; Acetyl-H3 Lys27; Acetyl-Histone H3 K27; Acetyl-Histone H3 Lys27; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K27ac; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; H3.3A; H3.3b; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-53; H3-614; H3a; H3b; H3-B; H3c1; H3c10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3f; H3-F; H3F1K; H3F2; H3F3; H3F3A; H3f3b; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3g; H3h; H3i; H3k27; H3Lys27ac; Hist1; Hist1h3a; HIST1H3B; Hist1h3c; HIST1H3D; Hist1h3e; HIST1H3F; HIST1H3G; HIST1H3H; Hist1h3i; HIST1H3J; HIST2H3A; Hist2h3b; HIST2H3C; Hist2h3c1; Hist2h3c2; Hist2h3c2-ps; Hist2h3ca1; Hist2h3ca2; HIST2H3D; histone 1, H3a; histone 1, H3b; histone 1, H3f; histone 1, H3h; histone 2, H3a; histone 2, H3c; histone 2, H3c2; histone 2, H3ca2; Histone 3; histone cluster 1, H3a; histone cluster 1, H3b; histone cluster 1, H3f; histone cluster 1, H3h; histone cluster 2, H3a; histone cluster 2, H3c; histone cluster 2, H3c2; histone cluster 2, H3c2, pseudogene; histone gene complex 1; histone H3; Histone H3.1; Histone H3.2; Histone H3.3; Histone H3.3C; Histone H3.5; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l; Histone H3/m; Histone H3/o; Histone H3K27; histone variant H3.5; M32461; PP781
H3-3A, H3-3B, H3-5, H3C1, H3C10, H3C14, H3C15, H3C2, H3C7
IgG
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.