Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3K27me1 Monoclonal Antibody (MABI 0321), Active Motif™

Mouse Monoclonal Antibody
$367.53 - $638.65
Specifications
Antigen | Histone H3K27me1 |
---|---|
Clone | MABI 0321 |
Concentration | 1 mg/mL |
Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3079
![]() ![]() |
Active Motif
61016 |
50 μg |
Each for $367.53
|
|
|||||
50-199-3078
![]() ![]() |
Active Motif
61015 |
100 μg |
Each for $638.65
|
|
|||||
Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Specifications
Histone H3K27me1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K27; H3.2; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-3A; H3-3B; H3a; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3F1K; H3F2; H3F3; H3F3A; H3F3B; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3k27; H3K27me; H3Lys27me; H3Lys27me1; HIST1H3A; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; HIST2H3A; HIST2H3C; HIST2H3D; histone 1, H3a; histone 1, H3b; histone 1, H3f; histone 1, H3h; histone 2, H3a; histone 2, H3c; Histone 3; histone cluster 1, H3a; histone cluster 1, H3b; histone cluster 1, H3f; histone cluster 1, H3h; histone cluster 2, H3a; histone cluster 2, H3c; Hist | |
H3-3A, H3-3B, H3-5, H3C1, H3C10, H3C14, H3C15, H3C2, H3C7 | |
IgG2a | |
Protein G | |
Antibody |
MABI 0321 | |
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P68431, P84243, Q6NXT2, Q71DI3 | |
126961, 3020, 3021, 333932, 440093, 8350, 8357, 8358, 8968 | |
This Histone H3 monomethyl Lys27 antibody was raised against a peptide containing monomethyl Lys27 of human Histone H3. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
H3-3A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title