Learn More
Histone H3K27me2me3 Monoclonal Antibody (7B11), Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H3K27me2me3 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 7B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM sodium phosphate with 150mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Host Species | Mouse |
| Immunogen | This Histone H3 di/trimethyl Lys27 antibody was raised against a peptide including trimethyl-lysine 27 of histone H3. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.