Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3K9ac Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992959
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2959 100 μg
50-199-2960 10 μg
50-227-7221 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-2959 Supplier Active Motif Supplier No. 39917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specifications

Antigen Histone H3K9ac
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene H3-3A
Gene Accession No. P02301, P68431, P68433, P84228, P84243, Q6NXT2, Q71DI3
Gene Alias Acetyl H3; Acetyl Histone H3; Acetyl-H3 K9; Acetyl-H3 Lys9; Acetyl-Histone H3 K9; Acetyl-Histone H3 Lys9; B0035.7; BUR5; CELE_T10C6.12; CG31613; CG31613 gene product from transcript CG31613-RA; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K9ac; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; H3.3A; H3.3b; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-1
Gene Symbols H3-3A, H3-3B, H3-5, H3C1, H3C10, H3C14, H3C15, H3C2, H3C7
Host Species Rabbit
Immunogen This Histone H3 acetyl Lys9 antibody was raised against a peptide including acetyl-lysine 9 of histone H3.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126961, 260423, 3020, 3021, 319150, 319152, 333932, 360198, 440093, 625328, 8350, 8357, 8358, 8968, 97114
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.