Learn More
Histone H3Kme3Q5Ser Rabbit anti-Human,Mouse, KHL, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Histone H3Kme3Q5Ser |
| Applications | ChIP Assay, Dot Blot, Western Blot |
| Classification | Polyclonal |
| Conjugate | KLH |
| Gene Accession No. | NP_003484 |
| Gene Alias | H3KmeQ5Ser |
| Gene Symbols | HIST3H3, H3FT |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3 trimethylated on lysine 4 and serotonylated on glutamine 5. |
| Molecular Weight of Antigen | ∼16 kDa observed; 15.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.