Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3Q5ser Rabbit anti-Human,Mouse, KHL, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$191.36 - $481.52
Specifications
Antigen | Histone H3Q5ser |
---|---|
Applications | ChIP Assay, Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | KLH |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE179125MI
|
MilliporeSigma
ABE179125UL |
25 μL |
Each for $191.36
|
|
|||||
ABE179110MI
|
MilliporeSigma
ABE1791100UL |
100 μL |
Each for $481.52
|
|
|||||
Description
Histone H3Q5ser Polyclonal antibody specifically detects Histone H3Q5ser in Human,Mouse samples. It is validated for ChIP-seq, Dot Blot, Western Blotting.Specifications
Histone H3Q5ser | |
Polyclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_003484 | |
HIST3H3, H3FT | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects Histone H3.1 serotonylated at glutamine 5 position. |
ChIP Assay, Dot Blot, Western Blot | |
KLH | |
Rabbit | |
Human, Mouse | |
H3Q5Ser | |
KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3.1 serotonylated on glutamine 5. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼16 kDa observed; 15.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title