Learn More
Histone H3R17me2aK18ac Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H3R17me2aK18ac |
| Applications | ChIP Assay, Dot Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | H3F3B |
| Gene Accession No. | P84244 |
| Gene Symbols | H3F3A, H3F3B |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.