Learn More
Histone H3S10ph Monoclonal Antibody (6G8B7), Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H3S10ph |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 6G8B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Host Species | Rat |
| Immunogen | This antibody was raised against a peptide corresponding to amino acids 1-19 including phospho-serine 10 of human Histone H3. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.