Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3S28ph Rat anti-Human, Mouse, Rat, Clone: HTA28, Active Motif™
SDP

Catalog No. 501994149
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-4149 Supplier Active Motif Supplier No. 61325
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specifications

Antigen Histone H3S28ph
Applications Western Blot
Classification Monoclonal
Clone HTA28
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.035% sodium azide; pH
Gene H3F3A
Gene Accession No. P84244
Gene Symbols H3F3A, H3F3B, H3F3C, HIST1H3B, Hist2h3c2
Host Species Rat
Immunogen This Histone H3 phospho Ser28 antibody was raised in rat against a peptide corresponding to amino acid residues 23-35 (phospho Ser 28) of human Histone H3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360868, 15078, 15081, 289314, 310678, 314977
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.