Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4, ac Lys12 Antibody (RM202), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326016MFV450
Description
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), Multiplex ImmunoassaySpecifications
Histone H4 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Vertebrates | |
Purified |
Western Blot, ELISA, Immunocytochemistry, ChIP Assay, Multiplex | |
RM202 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Chromatin Immunoprecipitation (ChIP), Multiplex Immunoassay | |
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
An acetyl-peptide corresponding to Histone H4 [ac Lys12] | |
0.1 mL | |
Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific | |
121504 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction