Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4, ac Lys16 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB212077F
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Porcine, Mammal samples. It is validated for Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (Negative).Specifications
Histone H4 (ac Lys16) | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (Negative) | |
P62805 | |
HIST4H4 | |
Synthetic acetylated peptide surrounding Lysine 16 of human Histone H4 [Swiss Prot P62805]. | |
0.05 mL | |
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones | |
121504 | |
Human, Mouse, Rat, Pig, Mammalia | |
IgG |
ChIP Assay, Immunofluorescence | |
FITC | |
PBS with 0.05% Sodium Azide | |
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse and rat. Predicted to react with most mammal species. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction