Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Histone H4, ac Lys5 Antibody, DyLight 650, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB212024C
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Porcine, Mammal samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Histone H4 (ac Lys5) | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| P62805 | |
| HIST4H4 | |
| Synthetic aceylated peptide surrounding Lysine 5 of human Histone H4 [Swiss Prot P62805]. | |
| 0.05 mL | |
| Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones | |
| 121504 | |
| Human, Mouse, Rat, Pig, Mammalia | |
| IgG | 
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse and rat. Predicted to react with most mammal species. | |
| Store at 4C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            