Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Histone H4, Dimethyl Lys44, Peptide
SDP

Catalog No. NBP19468010 Shop All R&D Systems Products
Change view
Click to view available options
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP19468010 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19468010 Supplier Novus Biologicals™ Supplier No. NBP1946801.0MG

Highly purified. Generating reliable and reproducible results.

The Histone H4 [Dimethyl Lys44] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.

Specifications

Gene ID (Entrez) 121504
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Formulation Lyophilized. Reconstitute in diH20.
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Product Type Histone H4
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Ac-RLARRGGV[K-Me2]RISGLIYE-Ahx-C-amide
Termination Unlabeled
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.