Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4, Monomethyl Lys20 Antibody (5E10-D8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130091AF594
Description
Histone H4 Monoclonal specifically detects Histone H4 in Human, Mouse, Rat, C. elegans, Drosophila, Mammal, Yeast samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Histone H4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
HIST4H4 | |
Branched peptide found between amino acids 10-100 of Histone H4. [UniProtNo. P62805] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, C. elegans, Drosophila, Mammalia, Yeast | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
5E10-D8 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
Mouse | |
Protein G purified | |
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones, Phospho Specific | |
121504.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction