Learn More
Histone H4, Monomethyl Lys20 Antibody (5E10-D8), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP130091IR
Catalog No. NBP130091IR
Only null left
Add to Cart
Description
Description
Histone H4 Monoclonal specifically detects Histone H4 in Human, Mouse, Rat, C. elegans, Drosophila, Mammal, Yeast samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Histone H4 (Monomethyl Lys20)
Monoclonal
DyLight 755
P62805
HIST4H4
Branched peptide found between amino acids 10-100 of Histone H4. [UniProt# P62805]
Protein G purified
RUO
Primary
All mammals including human, mouse and rat, Drosophila, C. elegans, yeast.
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
5E10-D8
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Mouse
11 kDa
0.1 mL
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
121504
Human, Mouse, Rat, C. elegans, Drosophila, Mammalia, Yeast
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only