Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4K16ac Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993936
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3936 10 μL
50-199-3934 50 μL
50-199-3935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-3936 Supplier Active Motif Supplier No. 39168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

Specifications

Antigen Histone H4K16ac
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene H4C1
Gene Accession No. P62805, P62806
Gene Alias acetyl Histone H4; Acetyl-H4 K16; Acetyl-H4 Lys16; Acetyl-Histone H4 K16; Acetyl-Histone H4 Lys16; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K16ac; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C2; H4c3; H4c4; H4C5; H4C6; H4C8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K16; H4K16a; H4Lys16ac; H4M; HIST1H4; HIST1H4A; Hist1h4b; Hist1h4c; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; Hist2h4a; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H4; Histone H4 acetyl; Histone H4A; Histone H4K16; Histone H4K16ac; histone IV, family 2; X04652
Gene Symbols H4-16, H4C1, H4C14, H4C2, H4C5, H4C9, His4r
Host Species Rabbit
Immunogen This Histone H4 acetyl Lys16 antibody was raised against a peptide including acetyl-lysine 16 of histone H4.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121504, 319158, 326619, 326620, 41773, 8294, 8359, 8366, 8367, 8370, 97122
Target Species Drosophila, Human, Mouse, Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.