Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4K20me3 Monoclonal Antibody (6F8-D9), Active Motif™
SDP

Catalog No. 501992773
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2773 10 μg
50-199-2772 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2773 Supplier Active Motif Supplier No. 39672
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

Specifications

Antigen Histone H4K20me3
Applications ChIP Assay, Dot Blot, Western Blot
Classification Monoclonal
Clone 6F8-D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.035% sodium azide; pH 7.5
Gene H4C1
Gene Accession No. P62805, P62806
Gene Alias CELE_T10C6.11; F07B7.11; F07B7.4; F45F2.12; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K20; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4c1; H4C11; H4c12; H4C13; H4C14; H4C15; H4C2; H4C3; H4c4; H4C5; H4C6; H4C8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K20; H4Lys20me3; H4M; his-20; his-22; his-4; his-52; his-54; his-8; HIST1H4; HIST1H4A; Hist1h4b; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; HIST2H4; Hist2h4a; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H2B 2; Histone H4; Histone H4A; Histone H4K20me3; histone IV, family 2; K06C4.12; K06C4.4; methyl Histone 3; methyl Histone H3; T10C6.11; Trimethyl H4; Tri-methyl H4; Trimethyl Histone H4; Tri-methyl Histone H4; Tri-Methyl-H4 K20; Tri-Methyl-H4 Lys20; Tri-Methyl-Histone H4 K20; Tri-Methyl-Histone H4 Lys20; X04652
Gene Symbols H4-16, H4C1, H4C14, H4C2, H4C5, H4C9
Host Species Mouse
Immunogen This Histone H4 trimethyl Lys20 antibody (mAb) was raised against a peptide containing trimethyl Lys20 of Histone H4.
Purification Method Protein G
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121504, 319158, 326619, 326620, 8294, 8359, 8366, 8367, 8370, 97122
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.