Learn More
Histone H4K31me1 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H4K31me1 |
| Applications | Dot Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Host Species | Rabbit |
| Immunogen | This Histone H4 monomethyl Lys31 antibody was raised against a peptide containing monomethyl Lys31 of human histone H4. |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.