Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4K5ac Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$156.65 - $638.65

Specifications

Antigen Histone H4K5ac
Concentration Conc. Not Determined
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-4066
SDP
Encompass
View Documents
Active Motif
39584
10 μL
Each for $156.65
Only null left
Add to Cart
 
50-199-4065
SDP
Encompass
View Documents
Active Motif
39583
200 μL
Each for $638.65
Only null left
Add to Cart
 
Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications

Specifications

Histone H4K5ac
ChIP Assay, Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Mouse
P62805, P62806
121504, 319158, 326619, 326620, 8294, 8359, 8366, 8367, 8370, 97122
This Histone H4 acetyl Lys5 antibody was raised against a peptide including acetyl-lysine 5 of histone H4.
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 30% glycerol and 0.035% sodium azide; pH
acetyl Histone H4; Acetyl-H4 K5; Acetyl-H4 Lys5; Acetyl-Histone H4 K5; Acetyl-Histone H4 Lys5; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K5ac; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4C11; H4c12; H4C13; H4c14; H4C15; H4c2; H4C3; H4C4; H4C5; H4c6; H4c8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K5; H4Lys5ac; H4M; HIST1H4; HIST1H4A; HIST1H4B; Hist1h4c; Hist1h4d; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; Hist2h4a; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histon
H4-16, H4C1, H4C14, H4C2, H4C5, H4C9
IgG
-20°C, Avoid Freeze/Thaw Cycles
H4C1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.