Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4R3me2a (asymmetric) Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $651.90
Specifications
Antigen | Histone H4R3me2a (asymmetric) |
---|---|
Concentration | Lot-Specific |
Applications | Dot Blot, ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2795
![]() ![]() |
Active Motif
39706 |
10 μL |
Each for $159.90
|
|
|||||
50-199-2796
![]() ![]() |
Active Motif
39006 |
50 μL |
Each for $375.15
|
|
|||||
50-199-2794
![]() ![]() |
Active Motif
39705 |
100 μL |
Each for $651.90
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.Specifications
Histone H4R3me2a (asymmetric) | |
Dot Blot, ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P62805 | |
121504, 8294, 8359, 8366, 8367, 8370 | |
This Histone H4 dimethyl Arg3, asymmetric antibody was raised against a synthetic peptide containing dimethyl-Arg3 (asymmetric) of human histone H4. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
H4C1 |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
dimethyl H4; di-methyl H4; dimethyl Histone H4; di-methyl Histone H4; Di-Methyl-H4 Arg3; Di-Methyl-H4 R3; Di-Methyl-Histone H4 Arg3; Di-Methyl-Histone H4 R3; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 R3me2a; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4Arg3me2; H4Arg3me2a; H4Arg3me2s; H4c1; H4c11; H4c12; H4C13; H4c14; H4C15; H4C2; H4c3; H4C4; H4C5; H4C6; H4C8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4M; H4R3; H4R3me2; H4R3me2s; HIST1H4; HIST1H4A; Hist1h4b; Hist1h4c; Hist1h4d; HIST1H4E; HIST1H4F; Hist1h4h; HIST1H4I; HIST1H4J; Hist1h4k; HIST1H4L; Hist1h4m; Hist2h4; Hist2h4a; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H4; Histone H4A; Histone H4R3; Histone H4R3me2; Histone H4R3me2a; histone IV, family 2; methyl Histone 3; methyl Histone H3; X04652 | |
H4-16, H4C1, H4C14, H4C2, H4C5, H4C9 | |
IgG | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title