Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone macroH2A2 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Specifications
| Antigen | Histone macroH2A2 |
|---|---|
| Concentration | Lot-Specific |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2918
![]()
|
Active Motif
39874 |
10 μg | N/A | N/A | N/A | ||||
50-199-2917
![]()
|
Active Motif
39873 |
100 μg | N/A | N/A | N/A | ||||
Description
Variant histone H2A which replaces conventional H2A in a subset of nucleosomes where it represses transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in stable X chromosome inactivation.Specifications
| Histone macroH2A2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9P0M6 | |
| 55506 | |
| This macroH2A2 antibody was raised against a peptide corresponding to amino acids 162-372 of human macroH2A2. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| H2AFY2 |
| Lot-Specific | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
| Core histone macro-H2A.2; core histone macroH2A2.2; H2A histone family member Y2; H2A histone family, member Y2; H2afy2; H2afy3; Histone macroH2A2; histone mH2A2; MACROH2A2; mH2A2; RGD1561371 | |
| H2AFY2 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title