Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Nucleosome Core Antibody, Novus Biologicals™
SDP

Catalog No. p-7199455 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017814 0.025 mg
NB100745 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017814 Supplier Novus Biologicals Supplier No. NB1007450.025MG
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Histone Nucleosome Core Polyclonal specifically detects Histone Nucleosome Core in Bovine samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Histone Nucleosome Core
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 2.42 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation PBS with 0.1% Sodium Azide
Host Species Sheep
Immunogen Intact calf H-3 complexed to rRNA.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Test Specificity This is specific for Histone-Core. The antibody binds to H1, H2B, H3 and H4. There is no evidence to suggest that the antibody can bind to H2A.
Target Species Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.