Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 8/HDAC8 Antibody (JJ0845), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267837
Description
Histone Deacetylase 8/HDAC8 Monoclonal specifically detects Histone Deacetylase 8/HDAC8 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
HistoneDeacetylase8/HDAC8 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
EC 3.5.1.98, HD8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3 | |
Rabbit | |
Protein A purified | |
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific | |
55869 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
JJ0845 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
HDAC8 | |
Synthetic peptide within Human Histone Deacetylase 8/HDAC8 aa 51-100/ 377. (SwissProt: Q9BY41 Human) | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction