Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 (Trimethyl Lys79) Rabbit anti-Human, Mouse, Rat, Clone: SR42-06, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267343
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Histone H3 (Trimethyl Lys79) | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K79Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SR42-06 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
H3C14 | |
Synthetic peptide within Human Histone H3 aa 56-105/ 136(tri methyl K79) conjugated to Keyhole Limpet Haemocyanin (KLH). (SwissProt: P68431 Human; SwissProt: P84243 Human; SwissProt: Q16695 Human; SwissProt: Q6NXT2 Human; SwissProt: Q71DI3 Human; SwissProt: P68433 Mouse; SwissProt: Q6LED0 Rat) | |
Protein A purified | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
8290 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction