Learn More
Simport™ Scientific Histosette™ I Biopsy Processing/Embedding Cassette
Shop All Simport Scientific ProductsDescription
- Made of high density polymer, acetal
- Keeps specimens safely submerged in liquid
- Resistant to chemical action of most histological solvents
- 45° angle anterior writing area
- One-piece integral lid eliminates need for separate steel lids
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely
- Available in 11 colors
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Specifications
| Writing Face Angle | 45° |
| Closure Included | Yes |
| Color | Peach |
| Material | Acetal |
| Product Type | Biopsy Cassette |
For in vitro diagnostic (IVD) use only.