Learn More
Simport™ Scientific Histosette™ I Biopsy Processing/Embedding Cassette
Catalog No. 22045075 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-045-075 Orange
22-045-066 White
22-045-067 Pink
22-045-068 Green
22-045-069 Yellow
22-045-070 Blue
22-045-071 Peach
22-045-072 Tan
22-045-073 Gray
22-045-074 Lilac
22-045-076 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-045-075 Supplier Simport™ Scientific Supplier No. M49911
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to hold biopsy specimens during the embedding process. Histosette™ I Biopsy Processing/Embedding Cassette are suitable to be used with some models of cassette labeling instruments.
  • Made of high density polymer, acetal
  • Keeps specimens safely submerged in liquid
  • Resistant to chemical action of most histological solvents
  • 45° angle anterior writing area
  • One-piece integral lid eliminates need for separate steel lids
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Back mounted tab allows for an unobstructed anterior marking area
  • Open and close as often as necessary - cassettes will always relock securely
  • Available in 11 colors
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Closure Included Yes
Writing Face Angle 45°
Color Orange
Material Acetal
Product Type Biopsy Cassette
For in vitro diagnostic (IVD) use only.