Learn More
Simport™ Scientific Histosette™ II Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038062 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Gray
Lilac
Orange
Peach
Pink
Tan
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-062 Tan
22-038-061 Peach
22-038-063 Gray
22-038-065 Orange
22-038-066 Aqua
22-038-057 Pink
22-038-064 Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-038-062 Supplier Simport™ Scientific Supplier No. M4938T
Add to Cart
Add to Cart
Secure specimens during processing with Simport Histosette™ II Biopsy Processing/Embedding Cassettes in Quickload™ Stacks Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
  • In Quickload™ sleeve
  • 45° anterior writing area
  • Open and close as often as necessary - cassettes will always relock securely
  • Back mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Tan
Material Acetal