Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Shop All Simport Scientific ProductsDescription
- Made of acetal
- Keeps specimens safely submerged in liquid
- Resistant to the chemical action of most histological solvents
- 45° angle anterior writing area
- One-piece integral lid eliminates need for separate steel lids
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely
- Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Specifications
| Closure Included | Yes |
| Color | Orange |
| Material | Acetal |
| Product Type | Tissue Cassette |
For in vitro diagnostic (IVD) use only.