Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hitfinder plate no. 007 for HTS, 80 compounds, 1 micromol each, Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Packaging:
Amber Glass Bottle
Specifications
Specifications
Chemical Name or Material | Hitfinder plate no 007 for HTS, 80 compounds 1micromol each |
Packaging | Amber Glass Bottle |
Quantity | 1 Ea. |
Formula Weight | 0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction