Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ AVB Sepharose™ Columns
GSA_VA

Catalog No. 4520059 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
5 x 1 mL
Diameter (Metric) Inner:
7 mm
16 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (Metric) Inner
45-200-59 5 x 1 mL 7 mm
45-200-60 1 x 5 mL 16 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-200-59 Supplier Cytiva Supplier No. 28411211
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Enables purification of adeno associated viruses (AAV) in one step, with high purity and excellent yields. Cytiva HiTrap™ AVB Sepharose™ Columns are pre-packed with affinity chromatography medium (resin) for simple operation with a syringe, pump or chromatographic system.

AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV). It enables purification of adeno associated viruses in one step with high purity and excellent yields.
  • One step purification of adeno associated viruses
  • Affinity resin with high selectivity and low background binding
  • Large surface area to give enhanced capacity
  • Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system
AVB Sepharose High Performance has proven affinity for adeno associated viruses from subclasses 1, 2, 3, and 5.
  • It is expected to have affinity for most of the other adeno associated virus subclasses
  • The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 1 mL
For Use With (Application) One Step Purification of Adeno Associated Viruses
Stability Stable to all commonly used aqueous buffers
Storage Requirements Store prepacked column at 4°C to 30°C in 20% Ethanol and Media at 4°C to 8°C in 20% Ethanol.
Binding Capacity >10▿12 genecopies/mL Chromatography Mediμm
Product Type HiTrap AVB Sepharose HP Column
Diameter (Metric) Inner 7 mm
Flow Rate <150 cm/hr
Particle Size 24 to 44 μm
Height (Metric) Bed 25 mm
Material Polypropylene
Max. Pressure 70 psi (5 bar)
Matrix Cross-linked agarose
pH Range 3 to 10
Bed Volume (Metric) 1 mL
Ligand Type 14 kD recombinant protein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.