Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Benzamidine FF (high sub) Columns

Catalog No. 45000289 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5mL
2 x 1mL
5 x 1mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Diameter (Metric) Inner:
0.7cm
1.6cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate Diameter (Metric) Inner
45-000-289 2 x 1mL <4 mL/min. 0.7cm
45-000-290 1 x 5mL <20 mL/min. 1.6cm
45-000-288 5 x 1mL <4 mL/min. 0.7cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-000-289 Supplier Cytiva Supplier No. 17514302
Add to Cart
Add to Cart

Remove or purify serine proteases such as thrombin. Cytiva Life Sciences™ HiTrap™ Benzamidine FF (high sub) Columns are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub).

  • Prepacked with Benzamidine Sepharose™ 4 Fast Flow (high sub) media
  • Protein binding capacity ≥35mg trypsin/mL medium
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 x 1mL
Flow Rate <4 mL/min.
Particle Size 90 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 1mL
Product Type Benzamidine FF Column
Diameter (Metric) Inner 0.7cm
For Use With (Application) Removal or purification of Serine Proteases such as Thrombin
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type >12μM p-aminobenzamidine/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 2 to 8
Support Type 4% Highly Cross-linked Agarose
Sample Type Serine Proteases such as Thrombin
Content And Storage 2° to 8°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.